Learn More
Invitrogen™ Attune™ CytPix™ No-Wash No-Lyse Filter Kit
The Attune CytPix No-Wash No-Lyse Filter Kit enables configuration of the Attune CytPix Flow Cytometer with an additional side scatter channel off of the violet laser. This allows no-wash, no-lyse immunophenotyping of whole blood.
Brand:  Invitrogen™ A50920
Additional Details : Weight : 1.00000kg
Product Code. 17404683
  • 998.00€ / Each
Estimated Shipment: 10-06-2024
to see stock.
Description
Description
  • Enables configuration of the Attune CytPix Flow Cytometer with an additional side scatter channel off of the violet laser
  • The Attune CytPix cytometer is quickly and easily configured to collect violet side scatter by changing dichroic and emission blades
  • Kit allows for no-wash, no-lyse immunophenotyping of whole blood
  • Immunophenotyping of whole blood using traditional flow cytometry is typically performed with lysis of the red blood cells (RBCs) followed by a wash step, which can result in significant cell loss and damage
  • With the Attune CytPix Flow cytometer users can acquire data quickly, even from dilute samples, using either a fluorescent reagent as a trigger to eliminate red blood cells or a differential side scatter using both blue and violet scatter
  • This is all done in combination with bright field imaging to aid in sample analysis and quality monitoring
  • The Attune CytPix No-Wash No-Lyse Filter Kit enables the side scatter approach
Specifications
Specifications
Attune CytPix Flow Cytometer
• 1 Dichroic 415 LP filter
• 1 Emission 405/10+OD2 BP filter
Store at room temperature.
No-Wash No-Lyse Filter Kit
No-Wash No-Lyse Applications
Promotions
For Research Use Only, Not for use in diagnostic procedures