Learn More
3M™ Cashew Protein Rapid Kit L25CHW
Intended for screening for the presence of cashew proteins in clean-in-place (CIP) final rinse water, environmental swab samples, food ingredients and processed food products.
Brand: 3M™ 7100151336
445.74 EUR valid until 2024-05-31
Use promo code "21654" to get your promotional price.
Additional Details : Weight : 0.02900kg
Description
- Results visible at 11 ± 1 minute after application of the sample.
- Rapid, qualitative immunochromatographic lateral flow test cartridges designed for the detection of allergenic food proteins.
- Designed to detect both processed and unprocessed allergen proteins.
- Test can accommodate environmental swab, CIP rinse water and food samples.
Specifications
Microbiology Media | |
Allergen | |
25/Cs. |