Learn More
Kimble Chase™ Chromaflex™ Glass Barrel
Brand:  Kimble Chase™ 4208311510
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11823063
  • 371.00€ / Each
Estimated Shipment: 29-08-2024
to see stock.
Specifications
25 mm
1 Each
Glass
Barrel
150 mm
25 mm