missing translation for 'onlineSavingsMsg'
Learn More

Cone and Plate Viscosity Standard, 250.2 mPa.s (25.00°C), Paragon Scientific™

Paragon ISO 17025 / ISO 17034 certified cone and plate viscosity standards are specifically formulated for the paint and coatings industry, and represent the most comprehensive range of standards available for this application.

Brand:  Paragon Scientific 2162/6

Additional Details : Weight : 0.90000kg

Product Code. 15929929

  • 154.00€ / 500mL
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Kinematic viscosity certified in strict accordance with ASTM D2162 at 20°C, 23°C & 25°C
  • Kinematic viscosity certified in accordance with ASTM D341 at intermediate temperatures
  • Density, dynamic viscosity and kinematic viscosity quoted at all temperatures
  • Ensures full compliance to ASTM & IP test method protocol
  • Full Traceability to National Standards
  • 2 year shelf life
  • Tamper evident security packaging
  • Manufactured in the United kingdom
Specifications

Specifications

Glass Amber Bottle
Yes
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Nominal values quoted. Certified Values will vary from batch to batch.