Learn More
Invitrogen™ Custom Primer
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole.
Brand:  Invitrogen™ 10629061
Additional Details : Weight : 0.00100kg
Product Code. 10674583
  • 0.55€ / Each
Estimated Shipment: 14-05-2024
to see stock.
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
Specifications
Specifications
10 nmol