Learn More
R&L Enterprises™ Double Burette Clamp
Securely holds burettes in place during the construction of laboratory retort stands. R&L Enterprises™ Double Burette Clamps are suitable for burettes up to 100mL capacity.
Brand:  R&L Enterprises™ 3596
Additional Details : Weight : 0.35100kg
Product Code. 11749027
  • 36.30€ / Each
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
Description
Description
  • Diecast
  • Without rod
  • With a single plastic headed thumbscrew to secure the clamp to retort rods, vertically or horizontally
Specifications
Specifications
1 Ea.
Double Clamp
with a single Plastic headed thumbscrew to secure the clamp to retort rods
For use up to 100mL burette