Learn More
Fisherbrand™ Individual Stainless Steel Leaf Calibration Weights, Class E2
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Leaf Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
41.98€
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
15535089
|
Fisherbrand™
|
2 mg |
41.98€
1 unit |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
15575089
|
Fisherbrand™
|
50 mg |
41.98€
1 unit |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
15585089
|
Fisherbrand™
|
100 mg |
41.98€
1 unit |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15595089
|
Fisherbrand™
|
200 mg |
41.98€
1 unit |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
15545089
|
Fisherbrand™
|
5 mg |
41.98€
1 unit |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
15555089
|
Fisherbrand™
|
10 mg |
41.98€
1 unit |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
15565089
|
Fisherbrand™
|
20 mg |
41.98€
1 unit |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
15525089
|
Fisherbrand™
|
1 mg |
41.98€
1 unit |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15505099
|
Fisherbrand™
|
500 mg |
41.98€
1 unit |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
Description
- Class E2 calibration weight
- In compliance with OIML R111
Specifications
E2 | |
Stainless Steel |