Learn More
Hamilton™ One-Hub Assembled Tubing Assemblies
Providing a variety of connection solutions in the laboratory.
35.00€ - 44.00€
Product Code | Brand | Needle Gauge | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Length (Metric) | Price | Quantity & Availability | ||||
10394703
|
Hamilton™
90611 |
11 G | 304.8 mm |
41.00€
Each |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10106833
|
Hamilton™
90610 |
10 G | 304.8 mm |
35.00€
Each |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11543084
|
Hamilton™
90612 |
12 G | 304.8 mm |
41.00€
Each |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10170883
|
Hamilton™
90613 |
13 G | 304.8 mm |
35.00€
Each |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11583124
|
Hamilton™
90672 |
20 G | Custom |
44.00€
Each |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11573124
|
Hamilton™
90670 |
20 G | Custom |
44.00€
Each |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11563084
|
Hamilton™
90614 |
14 G | 304.8 mm |
41.00€
Each |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11563124
|
Hamilton™
90669 |
19 G | Custom |
43.00€
Each |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11593134
|
Hamilton™
90662 |
12 G | Custom |
44.00€
Each |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11593084
|
Hamilton™
90617 |
17 G | 304.8 mm |
41.00€
Each |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11503134
|
Hamilton™
90676 |
26 G | Custom |
44.00€
Each |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11523134
|
Hamilton™
90680 |
30 G | Custom |
44.00€
Each |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11503084
|
Hamilton™
90608 |
8 G | 304.8 mm |
41.00€
Each |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11533144
|
Hamilton™
90666 |
16 G | Custom |
43.00€
Each |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Fitting manufactured from CTFE
- Tubing manufactured from PTFE
- For female luers
- A variety of gauges and lengths available