Learn More
Robu™ Series 15 0 Porosity Filter Plates
Brand:  Robu™ 51500
Additional Details : Weight : 1.00000kg
Product Code. 15298746
  • 163.00€ / Each
Estimated Shipment: 28-06-2024
to see stock.
Specifications
1Pack
Filter Plate
10 mm
150 mm