Learn More
Robu™ Series 15 0 Porosity Filter Plates
Brand:  Robu™ 15250
Additional Details : Weight : 0.01000kg
Product Code. 12060412
  • 42.00€ / Pack of 10
Estimated Shipment: 27-06-2024
to see stock.
Specifications
1Pack
Filter Plate
3 mm
25 mm