Learn More
Robu™ Series 15 1 Porosity Filter Plates
Brand:  Robu™ 15121
Additional Details : Weight : 0.10000kg
Product Code. 10317781
  • 71.05€ / Each
Estimated Shipment: 05-06-2024
to see stock.
Specifications
1Pack
Filter Plate
8.5 mm
120 mm