missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ 24-Well, cellGrade Plus-Treated, Smooth-Walled Inserts - Flat-Bottom Microplate

Color:Clear; Sterility:Sterile; Lid:Without Lid

Brand:  BRAND™ 782876

318.75 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 15593773

  • 348.00€ / Pack of 48
Estimated Shipment: 20-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For manual and automated cell and tissue cultivation
  • Consists of two different cell culture plates and two versions of cell culture inserts, which can also be combined with one another
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Without Lid
Smooth-walled
8 μm
Insert
Polystyrene
Sterile
Flat Bottom (F-well)
PET
48/Pk.
Clear
6-well BRANDplates™ Microplates
24
cellGrade Plus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions