Learn More
3M™ Clean-Trace™ Surface Protein (Allergen) Test Swab ALLTEC60
Rapid, simple, and reliable solution to monitor the presence of allergens on surfaces to effectively manage cleaning.
Brand: 3M™ Clean-Trace™ 7000034528
297.67 EUR valid until 2024-05-31
Use promo code "21654" to get your promotional price.
Additional Details : CAS : 7732-18-5 Weight : 0.48500kg
Description
- Detects as little as 3 μg of protein on surfaces and in hard to reach areas.
- Provides a tool to help assess and improve hygienic performance.
- Visual reading of a color change indicates the level of cleanliness.
- Real time results allow immediate corrective action.
- Validated for a range of allergenic proteins, including egg, milk, gluten, soy, peanut, almond and buckwheat.
Specifications
Surface Hygiene | |
60/Cs. |
Safety and Handling
- 3M Clean-Trace Surface Protein (Allergen) Test Swab
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : copper sulphate pentahydrate, sodium hydroxide