Learn More
3M™ Clean-Trace™ Water Positive Control HWATP10
Designed to provide assurance that your 3M hygiene monitoring reagents and instruments are being stored and used correctly.
Brand:  3M™ Clean-Trace™ 7000086277
Additional Details : Weight : 0.34500kg
Product Code. 17273734
  • 140.98€ / Pack of 10
Estimated Shipment: 02-09-2024
to see stock.
Add to basket
Description
Description
  • Verifies the performance of reagents and instruments
  • Can be used as a quench test with 3M™ Clean-Trace™ Water ATP tests
Specifications
Specifications
30 ng