Learn More
Microspec™ Wrappastack™ Replacement Stand for Starter Kit
Accessory for use with Starter kit, Sleeves
Brand: Microspec™ K3
Additional Details : Weight : 0.28900kg
Description
- Allows Wrapastack sleeves to shrink under bottom of stack of ten plates
-
Spécification
Steel | |
Wrapstack Sleeves, Starter Kit | |
60 mm |
Petri Dish Stand | |
10 mm |