accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 100-75-4

IUPAC Name: 1-nitrosopiperidine
Synonyms: N-nitrosopiperidine
Molecular Weight (g/mol): 114.15
Molecular Formula: C5H10N2O
InChi Key: UWSDONTXWQOZFN-UHFFFAOYSA-N
SMILES: O=NN1CCCCC1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results