accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNClOHO

CAS RN 1000017-93-5

IUPAC Name: 5-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Synonyms: 5-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Molecular Weight (g/mol): 196.59
Molecular Formula: C8H5ClN2O2
InChi Key: TVUHAHMCORHJBN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN2C(C=CC=C2Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results