accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNBrNH2N

CAS RN 1000018-06-3

IUPAC Name: 5-bromo-1-methyl-1H-indazol-3-amine
Synonyms: 5-bromo-1-methylindazol-3-amine
Molecular Weight (g/mol): 226.08
Molecular Formula: C8H8BrN3
InChi Key: BKBSBRJIGMVBFM-UHFFFAOYSA-N
SMILES: CN1N=C(N)C2=CC(Br)=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results