accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10010-67-0

IUPAC Name: sodium 2-[4-(2-sulfoethyl)piperazin-1-yl]ethane-1-sulfonate
Synonyms: sodium pipes pipes monosodium salt pipes monosodium monosodium pipes
Molecular Weight (g/mol): 324.34
Molecular Formula: C8H17N2NaO6S2
InChi Key: OGGAIRCLBMGXCZ-UHFFFAOYSA-M
SMILES: [Na+].OS(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results