accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10022-70-5

IUPAC Name: sodium pentahydrate hypochlorite
Synonyms: sodium pentahydrate hypochlorite
Molecular Weight (g/mol): 164.51
Molecular Formula: ClH10NaO6
InChi Key: FHKLOBNGYGFRSF-UHFFFAOYSA-N
SMILES: O.O.O.O.O.[Na+].[O-]Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results