accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClTeClClCl

CAS RN 10026-07-0

IUPAC Name: tetrachloro-λ⁴-tellane
Synonyms: tellurium tetrachloride
Molecular Weight (g/mol): 269.4
Molecular Formula: Cl4Te
InChi Key: SWLJJEFSPJCUBD-UHFFFAOYSA-N
SMILES: Cl[Te](Cl)(Cl)Cl
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)