accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1003-14-1

IUPAC Name: 2-propyloxirane
Synonyms: 2-propyloxirane
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: SYURNNNQIFDVCA-UHFFFAOYNA-N
SMILES: CCCC1CO1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results