accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CO

CAS RN 1003-38-9

IUPAC Name: 2,5-dimethyloxolane
Synonyms: 2,5-dimethyltetrahydrofuran
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: OXMIDRBAFOEOQT-UHFFFAOYNA-N
SMILES: CC1CCC(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results