accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOOHOOOHOHHOHOHOHOHOHOHOOHH2OH2O

CAS RN 10030-67-8

IUPAC Name: 2-{[4-hydroxy-2,5-bis(hydroxymethyl)-2-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol dihydrate
Synonyms: 2-{[4-hydroxy-2,5-bis(hydroxymethyl)-2-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol dihydrate
Molecular Weight (g/mol): 540.47
Molecular Formula: C18H36O18
InChi Key: LNVIPYYEBMNJIL-UHFFFAOYNA-N
SMILES: O.O.OCC1OC(CO)(OC2OC(CO)C(O)C(O)C2O)C(OC2OC(CO)C(O)C(O)C2O)C1O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)