accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10030-73-6

IUPAC Name: (9E)-hexadec-9-enoic acid
Synonyms: trans-palmitoleic acid t-9-hexadecenoic acid (E)-9-hexadecenoicacid palmitelaidic acid trans-9-palmitoleic acid
Molecular Weight (g/mol): 254.41
Molecular Formula: C16H30O2
InChi Key: SECPZKHBENQXJG-BQYQJAHWSA-N
SMILES: CCCCCC\C=C\CCCCCCCC(O)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)