accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OH2OH2OH2OH2OEuONOOONOOONOO

CAS RN 10031-53-5

IUPAC Name: europium(3+) hexahydrate trinitrate
Synonyms: europium(3+) hexahydrate trinitrate
Molecular Weight (g/mol): 446.07
Molecular Formula: EuH12N3O15
InChi Key: JVYYYCWKSSSCEI-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Eu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)