CAS RN 1003316-10-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-[3-[[4-(Chloromethyl)benzoyl]amino][1,1'-biphenyl]-4-yl]carbamic Acid tert-Butyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-[3-[[4-(Chloromethyl)benzoyl]amino][1,1'-biphenyl]-4-yl]carbamic Acid tert-Butyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.