accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 100367-48-4

NH2HOF

CAS RN 100367-48-4

IUPAC Name: 5-amino-2-fluorophenol
Synonyms: 5-amino-2-fluorophenol
Molecular Weight (g/mol): 127.12
Molecular Formula: C6H6FNO
InChi Key: XQYICHLOAMMXKE-UHFFFAOYSA-N
SMILES: NC1=CC(O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results