accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1003845-06-4

IUPAC Name: (2-chloropyrimidin-5-yl)boronic acid
Synonyms: 2-chloropyrimidin-5-ylboronic acid
Molecular Weight (g/mol): 158.35
Molecular Formula: C4H4BClN2O2
InChi Key: YTCIHPTZKKWKKC-UHFFFAOYSA-N
SMILES: OB(O)C1=CN=C(Cl)N=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results