accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10048-13-2

H3COO(R)O(S)OHOO

CAS RN 10048-13-2

IUPAC Name: (3S,7R)-15-hydroxy-11-methoxy-6,8,20-trioxapentacyclo[10.8.0.0²,⁹.0³,⁷.0¹⁴,¹⁹]icosa-1,4,9,11,14,16,18-heptaen-13-one
Synonyms: sterigmatocystin ccris 561 sterigmatocystine sterigmatocystin (substance)
Molecular Weight (g/mol): 324.29
Molecular Formula: C18H12O6
InChi Key: UTSVPXMQSFGQTM-DCXZOGHSSA-N
SMILES: COC1=C2C(=O)C3=C(O)C=CC=C3OC2=C2[C@@H]3C=CO[C@@H]3OC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results