CAS RN 100488-87-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CV-6209, TRC
CAS: 100488-87-7 Molecular Formula: C34 H60 N3 O6 . Cl Molecular Weight (g/mol): 642.31 Synonym: CV 6209 IUPAC Name: [2-methoxy-3-(octadecylcarbamoyloxy)propyl] N-acetyl-N-[(1-ethylpyridin-1-ium-2-yl)methyl]carbamate;chloride SMILES: [Cl-].CCCCCCCCCCCCCCCCCCNC(=O)OCC(COC(=O)N(Cc1cccc[n+]1CC)C(=O)C)OC