CAS RN 1005-64-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(E)-1-Phenyl-1-butene, TRC
CAS: 1005-64-7 Molecular Formula: C10 H12 Molecular Weight (g/mol): 132.2 Synonym: (1E)-1-Buten-1-ylbenzene (ACI),1-Butene, 1-phenyl-, (E)- (8CI),Benzene, (1E)-1-butenyl- (9CI),Benzene, 1-butenyl-, (E)- (ZCI),(E)-1-Phenyl-1-butene,(E)-But-1-en-1-ylbenzene,(E)-But-1-en-1-ylbenzene,trans-1-Phenyl-1-butene,trans-β-Ethylstyrene,(E)-1-Phenyl-1-butene IUPAC Name: [(E)-but-1-enyl]benzene SMILES: CC\C=C\c1ccccc1