CAS RN 1005006-69-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(4R)-3-[(2R,3S)-3-Hydroxy-1-oxo-2-(2-propen-1-yl)-4-penten-1-yl]-4-(phenylmethyl)-2-oxazolidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(4R)-3-[(2R,3S)-3-Hydroxy-1-oxo-2-(2-propen-1-yl)-4-penten-1-yl]-4-(phenylmethyl)-2-oxazolidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.