accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 100523-96-4

NIBr

CAS RN 100523-96-4

IUPAC Name: 2-bromo-4-iodopyridine
Synonyms: 2-bromo-4-iodopyridine
Molecular Weight (g/mol): 283.89
Molecular Formula: C5H3BrIN
InChi Key: HPKRNLGLZYOVJS-UHFFFAOYSA-N
SMILES: BrC1=NC=CC(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results