accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FeFeFeFeOPOOOPOOOOPOOOPOOOOPOOOPOOO

CAS RN 10058-44-3

IUPAC Name: tetrairon(3+) tri(phosphonatooxy)phosphonate
Synonyms: tetrairon(3+) tridiphosphate
Molecular Weight (g/mol): 745.2
Molecular Formula: Fe4O21P6
InChi Key: CADNYOZXMIKYPR-UHFFFAOYSA-B
SMILES: [Fe+3].[Fe+3].[Fe+3].[Fe+3].[O-]P([O-])(=O)OP([O-])([O-])=O.[O-]P([O-])(=O)OP([O-])([O-])=O.[O-]P([O-])(=O)OP([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results