CAS RN 1006-21-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Pyridoxine Cyclic Ether Impurity Hydrochloride Salt, TRC
CAS: 1006-21-9 Molecular Formula: C8 H9 N O2 . Cl H Molecular Weight (g/mol): 187.62 Synonym: 6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol Hydrochloride,Pyridoxine Hydrochloride Imp. A (EP) as Hydrochloride IUPAC Name: 6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol;hydrochloride SMILES: Cl.Cc1ncc2COCc2c1O
2
Pyridoxine EP Impurity A as Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.