accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1006-23-1

NH2NNH2NOH2NN

CAS RN 1006-23-1

IUPAC Name: 5-nitrosopyrimidine-2,4,6-triamine
Synonyms: 5-nitrosopyrimidine-2,4,6-triamine
Molecular Weight (g/mol): 154.13
Molecular Formula: C4H6N6O
InChi Key: XLQQJSWJHHKLOK-UHFFFAOYSA-N
SMILES: NC1=NC(N)=C(N=O)C(N)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results