accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BHOOH

CAS RN 100622-34-2

IUPAC Name: (anthracen-9-yl)boronic acid
Synonyms: anthracen-9-ylboronic acid
Molecular Weight (g/mol): 222.05
Molecular Formula: C14H11BO2
InChi Key: VHHDLIWHHXBLBK-UHFFFAOYSA-N
SMILES: OB(O)C1=C2C=CC=CC2=CC2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)