accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NFFFNNSOOCl

CAS RN 1006441-36-6

IUPAC Name: 1-[5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-4-sulfonyl chloride
Synonyms: 1-[5-(trifluoromethyl)pyridin-2-yl]pyrazole-4-sulfonyl chloride
Molecular Weight (g/mol): 311.66
Molecular Formula: C9H5ClF3N3O2S
InChi Key: ZACLAYCACGRQPN-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(C=C1)N1C=C(C=N1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results