accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10068-07-2

IUPAC Name: methyl 3-oxo-2,3-dihydro-1,2-oxazole-5-carboxylate
Synonyms: methyl 3-oxo-2H-1,2-oxazole-5-carboxylate
Molecular Weight (g/mol): 143.1
Molecular Formula: C5H5NO4
InChi Key: BBFWUUBQSXVHHZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=O)NO1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)