accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10075-50-0

IUPAC Name: 5-bromo-1H-indole
Synonyms: 5-bromoindole
Molecular Weight (g/mol): 196.05
Molecular Formula: C8H6BrN
InChi Key: VXWVFZFZYXOBTA-UHFFFAOYSA-N
SMILES: BrC1=CC=C2NC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results