accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOClNNH3CSOOHNOHNNOH3COCH3N

CAS RN 100784-20-1

IUPAC Name: methyl 3-chloro-5-({[(4,6-dimethoxypyrimidin-2-yl)carbamoyl]amino}sulfonyl)-1-methyl-1H-pyrazole-4-carboxylate
Synonyms: methyl 3-chloro-5-{[(4,6-dimethoxypyrimidin-2-yl)carbamoyl]aminosulfonyl}-1-methylpyrazole-4-carboxylate
Molecular Weight (g/mol): 434.81
Molecular Formula: C13H15ClN6O7S
InChi Key: FMGZEUWROYGLAY-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N(C)N=C1Cl)S(=O)(=O)NC(=O)NC1=NC(OC)=CC(OC)=N1
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results