accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1008-65-7

OHNNO(E)

CAS RN 1008-65-7

IUPAC Name: 6-[(2E)-2,3-dihydro-1,3,4-oxadiazol-2-ylidene]cyclohexa-2,4-dien-1-one
Synonyms: fenadiazole (inn)
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: HKVNQGYCUGIYTA-SOFGYWHQSA-N
SMILES: O=C1C=CC=C\C1=C1\NN=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results