accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1008-88-4

N

CAS RN 1008-88-4

IUPAC Name: 3-phenylpyridine
Synonyms: 3-phenylpyridine
Molecular Weight (g/mol): 155.2
Molecular Formula: C11H9N
InChi Key: HJKGBRPNSJADMB-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)