accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1008-91-9

IUPAC Name: 1-(pyridin-4-yl)piperazine
Synonyms: 1-(pyridin-4-yl)piperazine
Molecular Weight (g/mol): 163.22
Molecular Formula: C9H13N3
InChi Key: OQZBAQXTXNIPRA-UHFFFAOYSA-N
SMILES: C1CN(CCN1)C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)