CAS RN 101-72-4
CAS RN 101-72-4
IUPAC Name:
N1-phenyl-N4-(propan-2-yl)benzene-1,4-diamine
Synonyms:
IPPD -isopropylaminodiphenylamine
Molecular Weight (g/mol):
226.32
Molecular Formula:
C15H18N2
InChi Key:
OUBMGJOQLXMSNT-UHFFFAOYSA-N
SMILES:
CC(C)NC1=CC=C(NC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
N-Isopropyl-N'-phenyl-p-phenylenediamine, 95%
CAS: 101-72-4 Molecular Formula: C15H18N2 Molecular Weight (g/mol): 226.323 MDL Number: MFCD00026348 InChI Key: OUBMGJOQLXMSNT-UHFFFAOYSA-N Synonym: ippd,n-isopropyl-n'-phenyl-p-phenylenediamine,n-isopropyl-n'-phenyl-1,4-phenylenediamine,cyzone,4-anilino-n-isopropylaniline,antigen 3c,antioxidant ip,santoflex ip,diaphen fp,cyzone ip PubChem CID: 7573 ChEBI: CHEBI:63569 IUPAC Name: 1-N-phenyl-4-N-propan-2-ylbenzene-1,4-diamine SMILES: CC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2