accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

N

CAS RN 101-82-6

IUPAC Name: 2-benzylpyridine
Synonyms: 2-benzylpyridine
Molecular Weight (g/mol): 169.23
Molecular Formula: C12H11N
InChi Key: PCFUWBOSXMKGIP-UHFFFAOYSA-N
SMILES: C(C1=CC=CC=C1)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results