accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 101-89-3

IUPAC Name: 2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]benzene-1-diazonium hydrogen sulfate
Synonyms: 2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]benzenediazonium hydrogen sulfate
Molecular Weight (g/mol): 334.35
Molecular Formula: C14H14N4O4S
InChi Key: PPTXFSQSISUDAU-UHFFFAOYSA-M
SMILES: OS([O-])(=O)=O.CC1=CC=CC=C1N=NC1=CC=C([N+]#N)C(C)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results