accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOO

CAS RN 1010-60-2

IUPAC Name: 2-chloro-1,4-dihydronaphthalene-1,4-dione
Synonyms: 2-chloro-1,4-naphthoquinone C10H5ClO2
Molecular Weight (g/mol): 192.6
Molecular Formula: C10H5ClO2
InChi Key: CCTJHVLTAJTPBV-UHFFFAOYSA-N
SMILES: ClC1=CC(=O)C2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results