accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3CH3H3CCH3

CAS RN 1012-72-2

IUPAC Name: 1,4-di-tert-butylbenzene
Synonyms: 1,4-di-tert-butylbenzene
Molecular Weight (g/mol): 190.33
Molecular Formula: C14H22
InChi Key: OOWNNCMFKFBNOF-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results